Praseodymtrichlorid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
298298-5G
SIAL298298-5GEA
209
EUR
SIAL298298-5G
Praseodymtrichlorid, Sigma-Aldrich®
Praseodymtrichlorid
Formel:
Cl₃Pr Molecular Weight: 247,27 g/mol Schmelzpunkt: 786 °C Dichte: 4,02 g/cm³ (25 °C) |
MDL:
MFCD00011176 CAS-Nummer: 10361-79-2 EINECS: 233-794-4 Merck Index: 13,07709 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...