Chloropentaammincobalt(III)chlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

298301-10G
SIAL298301-10GEA 92 EUR
SIAL298301-10G
Chloropentaammincobalt(III)chlorid, Sigma-Aldrich®
Formel: H₁₅Cl₃CoN₅
MDL: MFCD00064728
CAS-Nummer: 13859-51-3
EINECS: 237-594-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR