(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4R,5S)-4-Methyl-5-phenyl-2-oxazolidone

M-Clarity™ Qualitätsstufe = MQ100

298891-5G
SIAL298891-5GEA 101 EUR
SIAL298891-5G
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinon, Sigma-Aldrich®
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinon
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2000 g/mol
MDL: MFCD00010845
CAS-Nummer: 77943-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR