Palladium(II)trifluoracetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Trifluoressigsäure Palladium(II)salz
M-Clarity™ Qualitätsstufe = MQ200
299685-1G
299685-5G
SIAL299685-1GEA
126
EUR
SIAL299685-1G
SIAL299685-5G
Palladium(II)trifluoracetat, Sigma-Aldrich®
Palladium(II)trifluoracetat
Formel:
C₄F₆O₄Pd |
MDL:
MFCD00013204 CAS-Nummer: 42196-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...