Palladium(II)trifluoracetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Trifluoressigsäure Palladium(II)salz

M-Clarity™ Qualitätsstufe = MQ200

299685-1G 299685-5G
SIAL299685-1GEA 126 EUR
SIAL299685-1G SIAL299685-5G
Palladium(II)trifluoracetat, Sigma-Aldrich®
Palladium(II)trifluoracetat
Formel: C₄F₆O₄Pd
MDL: MFCD00013204
CAS-Nummer: 42196-31-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR