1-Fluor-3-iod-5-nitrobenzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

299782-1G
SIAL299782-1GEA 51 EUR
SIAL299782-1G
1-Fluor-3-iod-5-nitrobenzol, Sigma-Aldrich®
1-Fluoro-3-iodo-5-nitrobenzene
Formel: C₆H₃FINO₂
Molecular Weight: 267 g/mol
MDL: MFCD00007219
CAS-Nummer: 3819-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR