2-Hydroxyfluoren, Sigma-Aldrich®

Lieferant: Merck

299847-100MG
SIAL299847-100MGEA 170 EUR
SIAL299847-100MG
2-Hydroxyfluoren, Sigma-Aldrich®
2-Hydroxyfluoren
Formel: C₁₃H₁₀O
MDL: MFCD00010778
CAS-Nummer: 2443-58-5
EINECS: 219-478-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR