4-Fluorencarbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

299987-100MG
SIAL299987-100MGEA 43.4 EUR
SIAL299987-100MG
4-Fluorencarbonsäure, Sigma-Aldrich®
4-Fluorencarbonsäure
Formel: C₁₄H₁₀O₂
Molecular Weight: 210,23 g/mol
MDL: MFCD00001112
CAS-Nummer: 6954-55-8
EINECS: 230-138-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR