2,2,2-Trifluoracetylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

300004-800G
SIAL300004-800GEA 0 EUR
SIAL300004-800G
2,2,2-Trifluoracetylchlorid, Sigma-Aldrich®
2,2,2-Trifluoracetylchlorid
Formel: C₂ClF₃O
CAS-Nummer: 354-32-5
EINECS: 206-556-2
UN: 3057
ADR: 2,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR