2,7-Dibrom-9-fluorenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,7-Dibromo-9H-fluoren-9-one

M-Clarity™ Qualitätsstufe = MQ100

300098-5G
SIAL300098-5GEA 76.8 EUR
SIAL300098-5G
2,7-Dibrom-9-fluorenon, Sigma-Aldrich®
2,7-Dibrom-9-fluorenon
Formel: C₁₃H₆Br₂O
Molecular Weight: 338 g/mol
MDL: MFCD00010790
CAS-Nummer: 14348-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR