Methyl-2-brom-5-nitrobenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Brom-5-nitrobenzoesäuremethylester

300217-1G
SIAL300217-1GEA 26 EUR
SIAL300217-1G
Methyl-2-brom-5-nitrobenzoat, Sigma-Aldrich®
Methyl-2-brom-5-nitrobenzoat
Formel: C₈H₆BrNO₄
Molecular Weight: 260,04 g/mol
Schmelzpunkt: 82 °C
MDL: MFCD00010867
CAS-Nummer: 6942-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR