4-Amino-2,3,5,6-tetrafluorpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3,5,6-Tetrafluoropyridin-4-amine

M-Clarity™ Qualitätsstufe = MQ100

300624-1G
SIAL300624-1GEA 26.6 EUR
SIAL300624-1G
4-Amino-2,3,5,6-tetrafluorpyridin, Sigma-Aldrich®
4-Amino-2,3,5,6-tetrafluoropyridine
Formel: C₅H₂F₄N₂
Molecular Weight: 166,08 g/mol
MDL: MFCD00010855
CAS-Nummer: 1682-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR