trans-4-Chlor-3-nitrozimtsäure ≥97%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: trans-3-(4-Chlor-3-nitrophenyl)propensäure, trans-4-Chlor-3-nitrocinnamsäure

M-Clarity™ Qualitätsstufe = MQ100

300667-5G
SIAL300667-5GEA 59 EUR
SIAL300667-5G
trans-4-Chlor-3-nitrozimtsäure ≥97%, Sigma-Aldrich®
trans-4-Chlor-3-nitrozimtsäure
Formel: C₉H₆ClNO₄
Molecular Weight: 227,601 g/mol
Schmelzpunkt: 188…190 °C (Lit.)
MDL: MFCD00063311
CAS-Nummer: 20797-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR