Chlor(di-tert-butyl)phosphin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

301558-100G 301558-25G 301558-5G
SIAL301558-100GEA 903 EUR
SIAL301558-100G SIAL301558-25G SIAL301558-5G
Chlor(di-tert-butyl)phosphin, Sigma-Aldrich®
Chlor(di-tert-butyl)phosphin
Formel: C₈H₁₈ClP
MDL: MFCD00008815
CAS-Nummer: 13716-10-4
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR