9-Fluorenon-1-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 9-Oxofluoren-1-carbonsäure

M-Clarity™ Qualitätsstufe = MQ100

301663-1G
SIAL301663-1GEA 67 EUR
SIAL301663-1G
9-Fluorenon-1-carbonsäure, Sigma-Aldrich®
9-Fluorenon-1-carbonsäure
Formel: C₁₄H₈O₃
Molecular Weight: 224,22 g/mol
MDL: MFCD00011537
CAS-Nummer: 1573-92-8
EINECS: 216-396-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR