Phenyl-2-pyridylketonoxim, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Phenyl 2-Pyridyl Ketoxime
M-Clarity™ Qualitätsstufe = MQ100
301884-5G
SIAL301884-5GEA
63.7
EUR
SIAL301884-5G
Phenyl-2-pyridylketonoxim, Sigma-Aldrich®
Phenyl-2-pyridylketonoxim
Formel:
C₁₂H₁₀N₂O |
MDL:
MFCD00006310 CAS-Nummer: 1826-28-4 EINECS: 217-374-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...