(1R)-(-)-Campher-10-sulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (1R)-(-)-2-Oxobornan-10-sulfonylchlorid, (1R)-(-)-2-Oxobornan-10-sulfonsäurechlorid, L(-)-Campher-10-sulfonylchlorid, L(-)-Campher-10-sulfonsäurechlorid, (1R)-(-)-Campher-10-sulfonsäurechlorid

M-Clarity™ Qualitätsstufe = MQ200

301949-5G
SIAL301949-5GEA 32 EUR
SIAL301949-5G
(1R)-(-)-Campher-10-sulfonylchlorid, Sigma-Aldrich®
(1R)-(-)-Campher-10-sulfonylchlorid
Formel: C₁₀H₁₅ClO₃S
Molecular Weight: 250,75 g/mol
Schmelzpunkt: 64…67 °C
MDL: MFCD00064155
CAS-Nummer: 39262-22-1
UN: 3261
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR