(S)-(+)-1-Phenyl-1,2-ethandiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-(+)-α,β-Dihydroxyethylbenzol, (S)-(+)-(1,2-Dihydroxyethyl)benzol, (S)-(+)-Phenylethylenglycol, (S)-(+)-Phenylethan-1,2-diol

302155-1G
SIAL302155-1GEA 159 EUR
SIAL302155-1G
(S)-(+)-1-Phenyl-1,2-ethandiol, Sigma-Aldrich®
(S)-(+)-1-Phenyl-1,2-ethandiol
Formel: C₈H₁₀O₂
Molecular Weight: 138,17 g/mol
Siedepunkt: 272 °C (1013 hPa)
Schmelzpunkt: 67 °C
MDL: MFCD00066256
CAS-Nummer: 25779-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR