(±)-Indolin-2-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Indoline-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

302244-25G 302244-5G
SIAL302244-25GEA 110 EUR
SIAL302244-25G SIAL302244-5G
(±)-Indolin-2-carbonsäure, Sigma-Aldrich®
(±)-Indolin-2-carbonsäure
Formel: C₉H₉NO₂
Molecular Weight: 163.18 g/mol
MDL: MFCD00010635
CAS-Nummer: 78348-24-0
EINECS: 278-899-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR