Geranylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

302791-5G
SIAL302791-5GEA 92.2 EUR
SIAL302791-5G
Geranylchlorid, Sigma-Aldrich®
Geranylchlorid
Formel: C₁₀H₁₇Cl
MDL: MFCD00000983
CAS-Nummer: 5389-87-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR