2'-Hydroxy-6'-methoxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Hydroxy-6-methoxyphenyl)ethanone

303046-5G
SIAL303046-5GEA 101 EUR
SIAL303046-5G
2'-Hydroxy-6'-methoxyacetophenon, Sigma-Aldrich®
2'-Hydroxy-6'-methoxyacetophenon
Formel: C₉H₁₀O₃
Molecular Weight: 166.18 g/mol
MDL: MFCD00008732
CAS-Nummer: 703-23-1
EINECS: 211-872-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR