Methylpentafluorbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl Pentafluorobenzoate

303070-5G
SIAL303070-5GEA 100 EUR
SIAL303070-5G
Methylpentafluorbenzoat, Sigma-Aldrich®
Methylpentafluorbenzoat
Formel: C₈H₃F₅O₂
Molecular Weight: 226,1002 g/mol
MDL: MFCD00012172
CAS-Nummer: 36629-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR