4,5-Dichlor-3(2H)-pyridazinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,5-Dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one

M-Clarity™ Qualitätsstufe = MQ100

303097-25G
SIAL303097-25GEA 32 EUR
SIAL303097-25G
4,5-Dichlor-3(2H)-pyridazinon, Sigma-Aldrich®
4,5-Dichlor-3(2H)-pyridazinon
Formel: C₄H₂Cl₂N₂O
Molecular Weight: 164,98 g/mol
MDL: MFCD00051504
CAS-Nummer: 932-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR