(R)-(+)-β-Citronellol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-(+)-Citronellol
M-Clarity™ Qualitätsstufe = MQ100
303461-1G
SIAL303461-1GEA
135
EUR
SIAL303461-1G
(R)-(+)-β-Citronellol, Sigma-Aldrich®
(R)-(+)-β-Citronellol
Formel:
C₁₀H₂₀O Molecular Weight: 156,27 g/mol |
CAS-Nummer:
1117-61-9 EINECS: 214-250-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...