(S)-(-)-β-Citronellol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-(-)-Citronellol

MQ100

303488-1G 303488-5G
SIAL303488-1GEA 58.1 EUR
SIAL303488-1G SIAL303488-5G
(S)-(-)-β-Citronellol, Sigma-Aldrich®
(S)-(-)-β-Citronellol
Formel: C₁₀H₂₀O
Molecular Weight: 156.27 g/mol
Dichte: 0.855 g/cm³ (20 °C)
MDL: MFCD00063214
CAS-Nummer: 7540-51-4
EINECS: 231-415-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR