4-Bromtetrafluoranilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Bromo-2,3,5,6-tetrafluoroaniline

M-Clarity™ Qualitätsstufe = MQ200

303631-5G
SIAL303631-5GEA 71 EUR
SIAL303631-5G
4-Bromtetrafluoranilin, Sigma-Aldrich®
4-Bromtetrafluoranilin
Formel: C₆H₂BrF₄N
Molecular Weight: 243.9844 g/mol
MDL: MFCD00012144
CAS-Nummer: 1998-66-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR