3-Nitro-5-(trifluormethyl)anisol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Methoxy-5-nitrobenzotrifluoride

M-Clarity™ Qualitätsstufe = MQ100

303674-5G
SIAL303674-5GEA 45.1 EUR
SIAL303674-5G
3-Nitro-5-(trifluormethyl)anisol, Sigma-Aldrich®
3-Nitro-5-(trifluormethyl)anisol
Formel: C₈H₆F₃NO₃
Molecular Weight: 221,13 g/mol
MDL: MFCD00010446
CAS-Nummer: 328-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR