3-Butylpyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

303933-10G
SIAL303933-10GEA 418 EUR
SIAL303933-10G
3-Butylpyridin, Sigma-Aldrich®
3-Butylpyridine
Formel: C₉H₁₃N
Molecular Weight: 135,21 g/mol
MDL: MFCD00010709
CAS-Nummer: 539-32-2
EINECS: 208-715-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR