(2-Hydroxyethyl)triphenylphosphoniumbromid, Sigma-Aldrich®
Lieferant: Merck
304131-25G
SIAL304131-25GEA
59.9
EUR
SIAL304131-25G
(2-Hydroxyethyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(2-Hydroxyethyl)triphenylphosphonium bromide
Formel:
C₂₀H₂₀BrOP Molecular Weight: 387,26 g/mol Schmelzpunkt: 219…220 °C |
MDL:
MFCD00011901 CAS-Nummer: 7237-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...