Methyl-2-amino-5-chlorbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-amino-5-chlorobenzoate

M-Clarity™ Qualitätsstufe = MQ100

305103-1G
SIAL305103-1GEA 45.8 EUR
SIAL305103-1G
Methyl-2-amino-5-chlorbenzoat, Sigma-Aldrich®
Methyl-2-amino-5-chlorbenzoat
Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
MDL: MFCD00007837
CAS-Nummer: 5202-89-1
EINECS: 225-992-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR