Sodium hexametaphosphate ≥96%, kristallin +200 mesh, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Polyphosphate sodium salt, Sodium polyphosphate, Calgon, Phosphate glass, water soluble, SHMP

M-Clarity™ Qualitätsstufe = MQ200

305553-1KG
SIAL305553-1KGEA 69.1 EUR
SIAL305553-1KG
Sodium hexametaphosphate ≥96%, kristallin +200 mesh, Sigma-Aldrich®
Sodium hexametaphosphate
Formel: (NaO₃P)n
Dichte: 2,4 g/cm³ (20 °C)
MDL: MFCD00134118
CAS-Nummer: 68915-31-1
EINECS: 272-808-3
Merck Index: 12,08814

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR