2-Brom-5-chlorbenzotrifluorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Brom-4-chlor-2-(trifluormethyl)benzol, 2-Brom-5-chlorbenzotrifluorid

M-Clarity™ Qualitätsstufe = MQ100

306088-5G
SIAL306088-5GEA 52 EUR
SIAL306088-5G
2-Brom-5-chlorbenzotrifluorid, Sigma-Aldrich®
2-Brom-5-chlorbenzotrifluorid
Formel: C₇H₃BrClF₃
Molecular Weight: 259,45 g/mol
MDL: MFCD00010308
CAS-Nummer: 344-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR