2-(Chlormethyl)tetrahydropyran, Sigma-Aldrich®

Lieferant: Merck

306371-5G
SIAL306371-5GEA 51.4 EUR
SIAL306371-5G
2-(Chlormethyl)tetrahydropyran, Sigma-Aldrich®
2-(Chlormethyl)tetrahydropyran
Formel: C₆H₁₁ClO
Molecular Weight: 134,61 g/mol
MDL: MFCD00006622
CAS-Nummer: 18420-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR