trans-2-Decenal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-2-Decenal

30658-10ML
SIAL30658-10MLEA 61.2 EUR
SIAL30658-10ML
trans-2-Decenal, Sigma-Aldrich®
trans-2-Decenal
Formel: C₁₀H₁₈O
Lagertemperatur: Raumtemperatur
MDL: MFCD00014679
CAS-Nummer: 3913-81-3
EINECS: 223-474-2
UN: 3082
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR