Scandium(III)chlorid Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

307858-1G
SIAL307858-1GEA 176 EUR
SIAL307858-1G
Scandium(III)chlorid Hydrat, Sigma-Aldrich®
Scandium(III)chlorid Hydrat
Formel: Cl₃Sc·H₂O
Molecular Weight: 169,33 g/mol
CAS-Nummer: 25813-71-2
EINECS: 233-799-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR