2,6-Dimethyl-5-heptenal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dimethylhept-5-enal

M-Clarity™ Qualitätsstufe = MQ100

307963-25G
SIAL307963-25GEA 45.6 EUR
SIAL307963-25G
2,6-Dimethyl-5-heptenal, Sigma-Aldrich®
2,6-Dimethyl-5-heptenal
Formel: C₉H₁₆O
Molecular Weight: 140,23 g/mol
MDL: MFCD00006981
CAS-Nummer: 106-72-9
EINECS: 203-427-2
UN: 1989
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR