N,N-Dimethylhexylamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N,N-Dimethylhexan-1-amine
308102-25G
308102-5G
SIAL308102-25GEA
211
EUR
SIAL308102-25G
SIAL308102-5G
N,N-Dimethylhexylamin, Sigma-Aldrich®
N,N-Dimethylhexylamin
Formel:
C₈H₁₉N Molecular Weight: 129.25 g/mol |
MDL:
MFCD00009522 CAS-Nummer: 4385-04-0 EINECS: 224-491-8 UN: 2733 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...