N,N'-Dimethyl-1,3-propandiamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N1,N3-Dimethylpropane-1,3-diamine, N,N′-Dimethyl-1,3-propanediamine

M-Clarity™ Qualitätsstufe = MQ200

308110-5G
SIAL308110-5GEA 109 EUR
SIAL308110-5G
N,N'-Dimethyl-1,3-propandiamin, Sigma-Aldrich®
N,N'-Dimethyl-1,3-propandiamin
Formel: C₅H₁₄N₂
Molecular Weight: 102,18 g/mol
MDL: MFCD00008292
CAS-Nummer: 111-33-1
EINECS: 203-859-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR