N-Propylethylendiamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Propyl-1,2-diaminoethan, 1-Amino-2-(propylamino)ethan

308145-5G
SIAL308145-5GEA 43.8 EUR
SIAL308145-5G
N-Propylethylendiamin, Sigma-Aldrich®
N-Propylethylendiamin
Formel: C₅H₁₄N₂
Molecular Weight: 102,18 g/mol
Siedepunkt: 147…150 °C (1013 hPa)
Dichte: 0,829 g/cm³ (20 °C)
MDL: MFCD00008172
CAS-Nummer: 111-39-7
EINECS: 203-864-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR