N-Propyl-1,3-propandiamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

308153-5G
SIAL308153-5GEA 125 EUR
SIAL308153-5G
N-Propyl-1,3-propandiamin, Sigma-Aldrich®
N-Propyl-1,3-propandiamin
Formel: C₆H₁₆N₂
CAS-Nummer: 23764-31-0
EINECS: 245-869-9
UN: 2734
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR