1,3-Phenylendiisocyanat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

308234-5G
SIAL308234-5GEA 112 EUR
SIAL308234-5G
1,3-Phenylendiisocyanat, Sigma-Aldrich®
1,3-Phenylendiisocyanat
Formel: C₈H₄N₂O₂
MDL: MFCD00002018
CAS-Nummer: 123-61-5
EINECS: 204-637-7
UN: 2811
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR