5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-en ([2.2.2]Benzocryptand, Kryptofix® 222B) 50% in Toluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

308579-100MG
SIAL308579-100MGEA 80.2 EUR
SIAL308579-100MG
5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-en ([2.2.2]Benzocryptand, Kryptofix® 222B) 50% in Toluol, Sigma-Aldrich®
5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-en ([2.2.2]Benzocryptand, Kryptofix® 222B)
Formel: C₂₂H₃₆N₂O₆
CAS-Nummer: 31250-18-7
EINECS: 250-532-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR