1-Butyl-3-methylimidazoliummethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-n-Butyl-3-methylimidazolium methanesulphonate

30881-100G-F
SIAL30881-100G-FEA 273 EUR
SIAL30881-100G-F
1-Butyl-3-methylimidazoliummethansulfonat, Sigma-Aldrich®
1-Butyl-3-methylimidazolium methanesulphonate
Formel: C₉H₁₈N₂O₃S
Molecular Weight: 234,32 g/mol
MDL: MFCD06798173
CAS-Nummer: 342789-81-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR