Iron(III) perchlorate hydrate, gelbe Kristalle, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

309281-100G
SIAL309281-100GEA 63.7 EUR
SIAL309281-100G
Iron(III) perchlorate hydrate, gelbe Kristalle, Sigma-Aldrich®
Iron(III) perchlorate hydrate

Used as chemical actinometer based on photolysis of ferrioxalate in presence of polyoxometalate in aqueous solution.


Iron(III) perchlorate hydrate is a useful starting material for the synthesis of a tris-salicylate Fe(III) complex having bidentate chelating units and in photolysis of ferrioxalate in presence of polyoxometalate in aqueous solution. Also Useful as an oxidant for conducting polymer nanoparticles synthesised in ionic liquid by chemical polymerisation.


It is a Useful starting material for the synthesis of a tris-salicylate Fe(III) complex having bidentate chelating units which allow two coordination modes.

Formel: Cl₃FeH₂O₁₃
MDL: MFCD00149705
CAS-Nummer: 15201-61-3
EINECS: 236-908-0
UN: 1481
ADR: 5.1,II

Bestellen

Spezifikation Testergebnisse

ICP Major Analysis, Confirms Iron Component Confirmed
Titration by Na₂S₂O₃ 10.0 - 12.5%
Chloride Content ≤0.10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR