D(-)-Sorbit 70% 70, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
D(-)-Glucit, (2R,3R,4R,5S)-(-)-1,2,3,4,5,6-Hexanhexol, D(-)-Sorbitol, D(-)-Glucitol
309532-100ML
309532-500ML
SIAL309532-100MLEA
104
EUR
SIAL309532-100ML
SIAL309532-500ML
D(-)-Sorbit 70% 70, Sigma-Aldrich®
D(-)-Sorbit
Formel:
HOH₂C(CH(OH))₄CH₂OH Molecular Weight: 182.17 g/mol Siedepunkt: 295 Dichte: 1.3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004708 CAS-Nummer: 50-70-4 EINECS: 200-061-5 Merck Index: 13,08797 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...