(S)-(+)-Ethylmandelat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: L-(+)-Mandelic acid ethyl ester, (S)-Ethyl 2-hydroxy-2-phenylacetate, Ethyl L-(+)-Mandelate

M-Clarity™ Qualitätsstufe = MQ100

309974-5G
SIAL309974-5GEA 72 EUR
SIAL309974-5G
(S)-(+)-Ethylmandelat, Sigma-Aldrich®
(S)-(+)-Ethylmandelat
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
MDL: MFCD00064249
CAS-Nummer: 13704-09-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR