(R)-(-)-Ethylmandelat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: D(-)-Mandelsäureethylester, (R)-(-)-2-Hydroxy-2-phenylethansäureethylester, (R)-(-)-Mandelsäureethylester, (R)-(-)-2-Hydroxy-2-phenylessigsäureethylester, D(-)-Ethylglykolat, D(-)-2-Hydroxy-2-phenylessigsäureethylester, (R)-(-)-Ethylglykolat, Ethyl-(R)-(-)-glykolat, D(-)-Ethylmandelat, D-2-Hydroxy-2-phenylethansäureethylester, Ethyl-D(-)-glykolat

M-Clarity™ Qualitätsstufe = MQ100

309982-5G
SIAL309982-5GEA 81.2 EUR
SIAL309982-5G
(R)-(-)-Ethylmandelat, Sigma-Aldrich®
(R)-(-)-Ethylmandelat
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
MDL: MFCD00064248
CAS-Nummer: 10606-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR