D(+)-Glucose-[6-13C] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

310808-100MG
SIAL310808-100MGEA 466 EUR
SIAL310808-100MG
D(+)-Glucose-[6-13C] (99% ¹³C ), Sigma-Aldrich®
D(+)-Glucose-[6-13C]
Formel: C₆H₁₂O₆
MDL: MFCD00148979
CAS-Nummer: 106032-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR