α,α,α,2-Tetrafluor-p-toluylsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Fluoro-4-(trifluoromethyl)benzoic acid
M-Clarity™ Qualitätsstufe = MQ100
310867-5G
SIAL310867-5GEA
118
EUR
SIAL310867-5G
α,α,α,2-Tetrafluor-p-toluylsäure, Sigma-Aldrich®
α,α,α,2-Tetrafluor-p-toluylsäure
Formel:
C₈H₄F₄O₂ Molecular Weight: 208,11 g/mol Schmelzpunkt: 160 °C |
MDL:
MFCD00010322 CAS-Nummer: 115029-24-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...