Diethylammoniumbromid, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

310905-100G
SIAL310905-100GEA 63 EUR
SIAL310905-100G
Diethylammoniumbromid, Sigma-Aldrich®
Diethylammoniumbromid
Formel: C₄H₁₂BrN
Molecular Weight: 154,05 g/mol
MDL: MFCD00012498
CAS-Nummer: 6274-12-0
EINECS: 228-466-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR